Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SET-beta, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1738MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1738MI 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1738MI Supplier MilliporeSigma™ Supplier No. ABN1738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein SET (UniProt Q9EQU5; also known as I-2PP2A, Phosphatase 2A inhibitor I2PP2A, TAF-I, Template-activating factor I) is encoded by the Set gene (Gene ID 56086) in murine species. Protein SET (patient SE translocation) adopted its name when it was originally identified in a leukemia study in 1992. SET interacts with signaling pathways that promote normal cell growth and proliferation, as well as tumorigenesis and metastasis. SET protein inhibits PP2A by enhancing lyase activity of P450c17, a substrate for PP2A when it is Ser- and Thr-phosphorylated. Alternative splicings result in multiple SET isoforms, including SET-alpha (TAF-I alpha), SET-beta (TAF-I beta), where the N-terminal 37-amino acid sequence in Set-alpha is replaced with a different sequence of a shorter length in Set-beta. In addition, there exists a shorter spliced Set-beta isoform without the nuclear localization signal (NLS). Both Set-alpha and SET-beta are predominantly localized in the nucleus, but differ in their expression levels among various cell and tissue types. SET-beta is resides predominantly in the brain relative to Set-alpha, whereas the opposite pattern was observed in tissues such as the spleen. SET-beta functions have been described both in the nucleus and cytoplasm. The presence of a nuclear localization signal (NLS) and phosphorylation govern SET-beta isoforms cellular localization. Proteolytic cleavage of SET-beta at Lys176 by granzyme A or at Asn-175 by asparagine endopeptidase results in the release of GzmA-activated DNase (GAAD) from a multiprotein complex. Uninhibited in the absence of the full-length Set-beta, GAAD causes cell death by its DNA-nicking activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SET-beta
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene SET
Gene Accession No. Q9EQU5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a sequence near the N-terminus of mouse/rat SET-beta.
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001191804
Test Specificity This polyclonal antiserum targets sequence specific to the spliced isoform SET-beta (TAF-I beta) of human, murine, and rat species (UniProt Q01105-2, Q9EQU5-2, Q63945-2). The epitope sequence is not present in SET-alpha (TAF-I alpha) or any other human, murine, and rat protein SET spliced isoforms reported by UniProt.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.