Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SET (I2PP2A), NT, Mouse, Unlabeled, Clone: 10E7,

Catalog No. MABN1843 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1843 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1843 Supplier MilliporeSigma™ Supplier No. MABN1843
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein SET (UniProt Q01105; also known as HLA-DR-associated protein II, IGAAD, Inhibitor of granzyme A-activated DNase, PHAPII, I-2PP2A, Phosphatase 2A inhibitor I2PP2A, TAF-I, Template-activating factor I) is encoded by the SET gene (Gene ID 6418) in human. Protein SET (patient SE translocation ) adopted its name when it was originally identified in a leukemia study in 1992. SET interacts with signaling pathways that promote normal cell growth and proliferation, as well as tumorigenesis and metastasis. SET protein inhibits PP2A by enhancing lyase activity of P450c17, a substrate for PP2A when it is Ser- and Thr-phosphorylated. Alternative splicings result in multiple SET isoforms, including SET-alpha (TAF-I alpha), SET-beta (TAF-I beta), where the N-terminal 37-amino acid sequence in Set-alpha is replaced with a different sequence of a shorter length in Set-beta. In addition, there exists a shorter spliced Set- isoform without the nuclear localization signal (NLS). Both Set-alpha and Set-beta are predominantly localized in the nucleus, but differ in their expression levels among various cell and tissue types. Set-beta is predominant in the brain relative to Set-alpha, whereas the opposite pattern was observed in tissues such as the spleen. Set-beta functions have been described both in the nucleus and cytoplasm. The presence of a nuclear localization signal (NLS) and phosphorylation govern Set- isoforms cellular localization. Proteolytic cleavage of Set-beta at Lys176 by granzyme A or at Asn-175 by asparagine endopeptidase (AEP) results in the release of GzmA-activated DNase (GAAD) from a multiprotein complex. Uninhibited in the absence of the full-length Set-eta, GAAD causes cell death by its DNA-nicking activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SET (I2PP2A), NT
Applications Western Blot
Classification Monoclonal
Clone 10E7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SET
Gene Accession No. Q01105
Host Species Mouse
Immunogen Recombinant human SET (I2PP2A).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001116293
Test Specificity Clone 10E7 detected both full-length human SET (I2PP2A) and the N-terminal fragment I2NTF generated by Asn endopeptidase (AEP) cleavage (Basurto-Islas, G., et al. (2013). J. Biol. Chem. 288(24):17495-17507).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.