Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SET Recombinant Rabbit Monoclonal Antibody (8E8M9)
GREENER_CHOICE

Catalog No. PIMA535772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535772 Supplier Invitrogen™ Supplier No. MA535772
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ESFFTWFTDH SDAGADELGE VIKDDIWPNP LQYYLVPDMD DEEGEGEEDD DDDEEEEGLE DIDEEGDEDE GEEDEDDDEG EEGEEDEGED D.

SET is a multitasking protein, involved in apoptosis, transcription, nucleosome assembly and histone binding. Isoform 2 anti-apoptotic activity is mediated by inhibition of the GZMA-activated DNase, NME1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SET
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8E8M9
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SET
Gene Accession No. Q01105, Q63945, Q9EQU5
Gene Alias 2610030F17Rik; 2PP2A; 5730420M11Rik; AA407739; Ab1-115; HLA-DR-associated protein II; hypothetical protein LOC538444; I IGAAD; I2PP2A; I-2PP2A; IGAAD; inhibitor of granzyme A-activated DNase; inhibitor-2 of protein phosphatase-2A; IPP2A2; Liver regeneration-related protein LRRGR00002; OTTHUMP00000022286; PHAPII; Phosphatase 2A inhibitor I2PP2A; protein phosphatase 2A inhibitor 2; protein phosphatase type 2A inhibitor; protein SET; RP11-216B9.3; Set; SET nuclear oncogene; SET nuclear proto-oncogene; SET translocation; SET translocation (myeloid leukemia-associated); SET-like protein; StF-IT-1; TAF-1a; TAF-1b; TAF-I; TAF-IBETA; Template-activating factor I; Template-Activating Factor-I, chromatin remodelling factor
Gene Symbols SET
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-290 of human SET/TAF1 (Q01105).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307947, 56086, 6418
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.