Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SET Mouse anti-Human, Clone: PCRP-SET-1C6, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen SET
Clone PCRP-SET-1C6
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123148
SDP
View Documents
Novus Biologicals
NBP30783720UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

SET Monoclonal specifically detects SET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

SET
0.2 mg/ml
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
10 mM PBS with 0.05% BSA
6418
IgG1
Protein A or G purified
PCRP-SET-1C6
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
2PP2A, HLA-DR-associated protein II, I2PP2A, I-2PP2A, IGAAD, Inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, IPP2A2, PHAPIITAF-I, Phosphatase 2A inhibitor I2PP2A, protein phosphatase type 2A inhibitor, protein SET, SET nuclear oncogene, SET translocation (myeloid leukemia-associated), TAF-IBETA, Template-activating factor I, Template-Activating Factor-I, chromatin remodelling factor
Recombinant full-length human SET protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.