Learn More
Invitrogen™ SET Polyclonal Antibody

Description
PA1-29799 detects INHAT1 from human samples.
Specifications
Specifications
| Antigen | SET |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% BSA and 0.02% sodium azide |
| Gene | SET |
| Gene Accession No. | Q01105, Q63945, Q9EQU5 |
| Gene Alias | 2610030F17Rik; 2PP2A; 5730420M11Rik; AA407739; Ab1-115; HLA-DR-associated protein II; hypothetical protein LOC538444; I IGAAD; I2PP2A; I-2PP2A; IGAAD; inhibitor of granzyme A-activated DNase; inhibitor-2 of protein phosphatase-2A; IPP2A2; Liver regeneration-related protein LRRGR00002; OTTHUMP00000022286; PHAPII; Phosphatase 2A inhibitor I2PP2A; protein phosphatase 2A inhibitor 2; protein phosphatase type 2A inhibitor; protein SET; RP11-216B9.3; Set; SET nuclear oncogene; SET nuclear proto-oncogene; SET translocation; SET translocation (myeloid leukemia-associated); SET-like protein; StF-IT-1; TAF-1a; TAF-1b; TAF-I; TAF-IBETA; Template-activating factor I; Template-Activating Factor-I, chromatin remodelling factor |
| Gene Symbols | SET |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.