Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETBP1 Mouse anti-Mouse, Human, Clone: 63, MilliporeSigma™

Catalog No. MABE1128MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1128MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1128MI Supplier MilliporeSigma Supplier No. MABE1128
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SETBP1, clone 63 is a highly specific mouse monoclonal antibody that targets SETBP1 and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETBP1
Applications Western Blot
Classification Monoclonal
Clone 63
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SETBP1,KIAA0437,MRD29
Host Species Mouse
Immunogen GST-tagged recombinant human SETBP1 N-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001123582
Test Specificity Clone 63 targets the N-terminal region of human and mouse SETBP1.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.