Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561863 Supplier Invitrogen™ Supplier No. PA561863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDWVKNAQKA FDNTEGKREG YSADSAQEAS PARQNVSSAS NPENDSSHVR ITIPIKAPSL DPTNHKRKKR QSIKAVVEKI MPEKALASGI TMSSEVVNRI LSNSEGNKKD PRVPKLSKMI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

This gene encodes a protein which contains a several motifs including a ski homology region and a SET-binding region in addition to three nuclear localization signals. The encoded protein has been shown to bind the SET nuclear oncogene which is involved in DNA replication. Mutations in this gene are associated with Schinzel-Giedion midface retraction syndrome. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETBP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SETBP1
Gene Accession No. Q9Y6X0
Gene Alias C130092E12; Kiaa0437; mKIAA0437; MRD29; Seb; SET binding protein 1; SET-binding protein; SET-binding protein-like; SETBP1
Gene Symbols SETBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SETBP1. Recombinant protein control fragment (Product #RP-91649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26040
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.