Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD1A Antibody (OTI7B7), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274144AF750
Description
SETD1A Monoclonal specifically detects SETD1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SETD1A | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SETD1A | |
Recombinant protein fragment corresponding to amino acids 1-320 of human SETD1A (NP_055527) produced in E.coli. | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI7B7 | |
Western Blot, Immunohistochemistry | |
histone-lysine N-methyltransferase SETD1A, hSET1A, KIAA0339EC 2.1.1.43, KMT2FSET1, Lysine N-methyltransferase 2F, SET domain containing 1A, SET domain-containing protein 1A, Set1, Set1/Ash2 histone methyltransferase complex subunit SET1, SET1A | |
Mouse | |
0.1 mL | |
9739 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction