Learn More
Invitrogen™ SETD1A Polyclonal Antibody

Description
Immunogen sequence: SQFRSSDANY PAYYESWNRY QRHTSYPPRR ATREEPPGAP FAENTAERFP PSYTSYLPPE PSRPTDQDYR PPASEA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Specifications
Specifications
| Antigen | SETD1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SETD1A |
| Gene Accession No. | O15047 |
| Gene Alias | BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A |
| Gene Symbols | SETD1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.