Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578298 Supplier Invitrogen™ Supplier No. PA578298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SETD1A
Gene Accession No. O15047
Gene Alias BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A
Gene Symbols SETD1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SETD1A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233904, 9739
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.