Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116810 Supplier Invitrogen™ Supplier No. PA5116810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Jurkat, HepG2, U-251MG, Mouse testis Immunogen sequence: SSFGQFTPQS SQGTPYTSRG STPYSQDSAY SSSTTSTSFK PRRSENSYQD AFSRRHFSAS SASTTASTAI AATTAATASS SASSSSLSSS SSSSSSSSSS QFRSSDANYP AYYESWNRYQ RHTSYPPRRA TREEPPGAPF AENTAERFPP SYTSYLPPEP SRPTDQDYRP PASEAPPPEP PEPGGGGGGG GPSPEREEVR TSPRPASPAR SGSPAPETTN ESVPFAQHSS LDSRIEMLLK EQRSKFSFLA SDTEEEEENS SMVLGARDTG SEVPSGSGHG PCTPPPAPAN FEDVAPTGSG EPGATRESPK ANGQNQASPC SSGDDMEISD DDRGGSPPPA PTPPQQPPPP PPPPPPPPPY LASLPLGYPP HQPAYLLPPR PDGPPPPEYP PPPPPPPHIY DFVNSLELMD RLGAQWGGMP MSFQMQTQML TRLHQLRQGK GLIAASAGPP GGAFGEAF.

SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SETD1A
Gene Accession No. O15047
Gene Alias BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A
Gene Symbols SETD1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 258-458 of human SET1A (O15047).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233904, 9739
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.