Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD1A Recombinant Rabbit Monoclonal Antibody (BLR024E)
GREENER_CHOICE

Catalog No. PIMA544243
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544243 100 μL
PIMA544244 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544243 Supplier Invitrogen™ Supplier No. MA544243
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SETD1A Recombinant Monoclonal Antibody for Western Blot, IHC (P), IP

SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD1A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BLR024E
Concentration 10 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene SETD1A
Gene Accession No. O15047
Gene Alias BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A
Gene Symbols SETD1A
Host Species Rabbit
Immunogen Peptide representing a region between residues 1200 and 1250 of human hSET1 (KIAA0339).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233904, 9739
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.