Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554491 Supplier Thermo Scientific Supplier No. PA554491
Add to Cart
Add to Cart

SETD1B Polyclonal Antibody

SETD1B Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD1B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Setd1b
Gene Accession No. Q9UPS6
Gene Alias AA516740; BC035291; Histone-lysine N-methyltransferase SETD1B; hSET1B; Kiaa1076; KMT2G; LOC100359816; Lysine N-methyltransferase 2G; mKIAA1076; rCG21620-like; SET domain containing 1B; SET domain-containing protein 1B; SET1B; SETD1B
Gene Symbols Setd1b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SETD1B. Recombinant protein control fragment (Product #RP-93579).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23067
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.