Learn More
Invitrogen™ SETD1B Polyclonal Antibody

Description
Immunogen sequence: GLQFVNLPPYR GPFSLSNSGP GRGQHWPPLP KFDPSVPPPG YMPRQEDPHK ATVDGVLLVV LKELKAIMK Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
Specifications
Specifications
| Antigen | SETD1B |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Setd1b |
| Gene Accession No. | Q9UPS6 |
| Gene Alias | AA516740; BC035291; Histone-lysine N-methyltransferase SETD1B; hSET1B; Kiaa1076; KMT2G; LOC100359816; Lysine N-methyltransferase 2G; mKIAA1076; rCG21620-like; SET domain containing 1B; SET domain-containing protein 1B; SET1B; SETD1B |
| Gene Symbols | Setd1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.