Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102710
Description
Antibody detects endogenous levels of total SETD2.
Histone methyltransferase that specifically trimethylates 'Lys-36' of histone H3 (H3K36me3) using dimethylated 'Lys-36' (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintenance of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.
Specifications
SETD2 | |
Polyclonal | |
Unconjugated | |
Setd2 | |
4921524K10Rik; BC031601; FLJ16420; FLJ22472; FLJ23184; FLJ45883; HBP231; HIF1; HIF-1; HIP-1; histone-lysine N-methyltransferase SETD2; hSET2; HSPC069; huntingtin interacting protein 1; huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; HYPB; Kiaa1732; Kif9; kinesin family member 9; KMT3A; LLS; lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain containing 2; SET domain containing 2, histone lysine methyltransferase; SET domain-containing protein 2; SET2; Setd2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
235626, 29072, 316013 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
E9Q5F9, Q9BYW2 | |
Setd2 | |
A synthesized peptide derived from human SETD2(Accession Q9BYW2), corresponding to amino acid residues R507-S557. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction