Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534935 Supplier Invitrogen™ Supplier No. PA534935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat. Predicted reactivity: Rat (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Histone methyltransferase that specifically trimethylates 'Lys-36' of histone H3 (H3K36me3) using dimethylated 'Lys-36' (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintenance of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Setd2
Gene Accession No. Q9BYW2
Gene Alias 4921524K10Rik; BC031601; FLJ16420; FLJ22472; FLJ23184; FLJ45883; HBP231; HIF1; HIF-1; HIP-1; histone-lysine N-methyltransferase SETD2; hSET2; HSPC069; huntingtin interacting protein 1; huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; HYPB; Kiaa1732; Kif9; kinesin family member 9; KMT3A; LLS; lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain containing 2; SET domain containing 2, histone lysine methyltransferase; SET domain-containing protein 2; SET2; Setd2
Gene Symbols Setd2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human HYPB/SETD2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.