Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588104 Supplier Invitrogen™ Supplier No. PA588104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNRPDSRLGK TELSFSSSCE IPHVDGLHSS EELRNLGWDF SQEKPSTTYQ QPDSSYGACG GHKYQQNAEQ YGGTRDYWQG NGYWDPRSGR PPGTGVVYDR TQGQVPDSLT DDREEEENWD QQDGSHFSDQ SDKFLLSLQK DKGSVQAPEI SSNSIKDTLA VNEKKDFSKN LEKNDIKDRG PLKKRRQEIE SDSESDGELQ DRKKVRVE Positive Samples: Mouse brain, Rat brain; Cellular Location: Chromosome, Nucleus.

Histone methyltransferase that specifically trimethylates 'Lys-36' of histone H3 (H3K36me3) using dimethylated 'Lys-36' (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintenance of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Setd2
Gene Accession No. E9Q5F9
Gene Alias 4921524K10Rik; BC031601; FLJ16420; FLJ22472; FLJ23184; FLJ45883; HBP231; HIF1; HIF-1; HIP-1; histone-lysine N-methyltransferase SETD2; hSET2; HSPC069; huntingtin interacting protein 1; huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; HYPB; Kiaa1732; Kif9; kinesin family member 9; KMT3A; LLS; lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain containing 2; SET domain containing 2, histone lysine methyltransferase; SET domain-containing protein 2; SET2; Setd2
Gene Symbols Setd2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235626, 316013
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.