Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SETD3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SETD3 Polyclonal specifically detects SETD3 in Human samples. It is validated for Western Blot.Specifications
| SETD3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C14orf154, chromosome 14 open reading frame 154, DKFZp761E1415, EC 2.1.1.43, FLJ23027, MGC87236, SET domain containing 3, SET domain-containing protein 3 | |
| SETD3 | |
| IgG | |
| 34 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_954574 | |
| 84193 | |
| Synthetic peptide directed towards the middle region of human SETD3The immunogen for this antibody is SETD3. Peptide sequence AVSSVMTRQNQIPTEDGSRVTLALIPLWDMCNHTNGLTPEDSFALAVASA. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title