Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543675
Description
Peptide sequence: AVSSVMTRQN QIPTEDGSRV TLALIPLWDM CNHTNGLTPE DSFALAVASA Sequence homology: Human: 100%.
SETD3 is a histone methyltransferase that is involved in epigenetic transcriptional activation by methylating lysine residues on histone H3 tails.
Specifications
SETD3 | |
Polyclonal | |
Unconjugated | |
SETD3 | |
2610102I01Rik; 2610305M23Rik; 4921524K10Rik; 6030490B17Rik; Actin-histidine N-methyltransferase; BC031601; C14orf154; D12Ertd771e; Endothelial differentiation inhibitory protein D10; histone-lysine N-methyltransferase SETD2; histone-lysine N-methyltransferase setd3; hSETD3; Kiaa1732; KMT3A; Lysine N-methyltransferase 3A; Protein-lysine N-methyltransferase SETD2; RGD1309550; SET domain containing 2; SET domain containing 3; SET domain-containing protein 2; SET domain-containing protein 3; Setd2; Setd3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84193 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86TU7 | |
SETD3 | |
Synthetic peptide directed towards the middle region of human SETD3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction