Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582105 Supplier Invitrogen™ Supplier No. PA582105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERASPNSFWQ PYIQTLPSEY DTPLYFEEDE VRYLQSTQAI HDVFSQYKNT ARQYAYFYKV IQTHPHANKL PLKDSFTYED YRWAVSSVMT RQNQIPTEDG SR Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 97%.

SETD3 is a histone methyltransferase that is involved in epigenetic transcriptional activation by methylating lysine residues on histone H3 tails.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD3
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SETD3
Gene Accession No. Q86TU7, Q91WC0
Gene Alias 2610102I01Rik; 2610305M23Rik; 4921524K10Rik; 6030490B17Rik; Actin-histidine N-methyltransferase; BC031601; C14orf154; D12Ertd771e; Endothelial differentiation inhibitory protein D10; histone-lysine N-methyltransferase SETD2; histone-lysine N-methyltransferase setd3; hSETD3; Kiaa1732; KMT3A; Lysine N-methyltransferase 3A; Protein-lysine N-methyltransferase SETD2; RGD1309550; SET domain containing 2; SET domain containing 3; SET domain-containing protein 2; SET domain-containing protein 3; Setd2; Setd3
Gene Symbols SETD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SETD3. Recombinant protein control fragment (Product #RP-89073).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299295, 52690, 84193
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.