Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596629 Supplier Invitrogen™ Supplier No. PA596629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSAPQNPPQR KKVSLLEYRK RKQEAKENSA GGGGDSAQSK SKSAGAGQGS SNSVSDTGAH GVQGSSARTP SSPHKKFSPS HSSMSHLEAV SPSDSRGTSS Positive Samples: MCF7.

This function of this gene has yet to be determined but mutations in this gene have been associated with autosomal dominant mental retardation-23. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SETD5
Gene Accession No. Q5XJV7, Q9C0A6
Gene Alias 2900045N06Rik; C330007C20; C85544; Histone-lysine N-methyltransferase SETD5; histone-lysine N-methyltransferase SETD5; SET domain-containing protein 5; Kiaa1757; LOW QUALITY PROTEIN: SET domain-containing protein 5; mKIAA1757; RGD1310433; SET domain containing 5; SET domain-containing protein 5; SETD5
Gene Symbols SETD5
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297514, 55209, 72895
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.