Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596615 Supplier Invitrogen™ Supplier No. PA596615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPYPDNTDDT ADIQMVTVRE AALQGTKTEA ERHLVYERWD FLCKLEMVGE EGAFVIGREE VLTEEELTTT LKVLCMPAEE FRELKDQDGG GDDKREEGSL TITNIPKLKA SWRQLLQNSV LLTLQTYATD LKTDQGLLSN KEVYAKLSWR EQQALQVRYG QKMILHQLLE LTS; Positive Samples: BT-474, Mouse liver, Mouse kidney, Rat brain, Rat liver; Cellular Location: Nucleus.

SETD6 (SET domain-containing protein 6) is a 473 amino acid protein that belongs to the SETD6 family. Containing one SET domain, SETD6 is structurally similar to the Rubisco large subunit methyltransferase. SETD6 is a proteinlysine N-methyltransferase that specifically monomethylates 'Lys-310' of the NFκB p65 subunit of NFκB complex, leading to down-regulate NFκB transcription factor activity. SETD6-mediated methylation renders NFκB p65 inert and attenuates NFκB p65-driven transcriptional programs, including inflammatory responses in primary immune cells. The SETD6-initiated lysine-methylation signaling cascade acts to restrain activation of NFκB-mediated inflammatory responses in diverse cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SETD6
Gene Accession No. D3ZSK5, Q8TBK2, Q9CWY3
Gene Alias 0610039J04Rik; 3110004G14Rik; AI413388; C76402; N-lysine methyltransferase SETD6; RGD1560538; SET domain containing 6; SET domain-containing protein 6; SETD6
Gene Symbols SETD6
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291844, 66083, 79918
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.