Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7/9 Antibody (001), DyLight 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289842G
Description
SETD7/9 Monoclonal antibody specifically detects SETD7/9 in Human samples. It is validated for ELISASpecifications
SETD7/9 | |
Monoclonal | |
DyLight 488 | |
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SETD7/9 (NP_085151.1; Asp2-Lys366). | |
0.1 mL | |
Histones and Modified Histones, Signal Transduction | |
80854 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
001 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction