Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7/9 Antibody (s4E5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104298H
Description
SETD7/9 Monoclonal specifically detects SETD7/9 in Human samples. It is validated for Western Blot, ELISA.Specifications
SETD7/9 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SETD7 | |
Recombinant human SET7/9 protein | |
0.1 mL | |
Histones and Modified Histones, Signal Transduction | |
80854 | |
Human | |
Purified |
Western Blot, ELISA | |
s4E5 | |
Western Blot, ELISA | |
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human SET7/9 mAb, clone s4E5, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with recombinant human SET7/9. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction