Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7/9 Antibody (s4E5), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104298PEATT594
Description
SETD7/9 Monoclonal specifically detects SETD7/9 in Human samples. It is validated for Flow Cytometry.Specifications
SETD7/9 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
SETD7 | |
Recombinant human SET7/9 (1-366aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human SET7/9 mAb, clone s4E5, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with recombinant human SET7/9. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
s4E5 | |
Flow Cytometry | |
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
Mouse | |
Protein G purified | |
Histones and Modified Histones, Signal Transduction | |
80854.0 | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction