Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD7 Monoclonal Antibody (7C1B7E1), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543017
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543017 100 μL
PIMA543500 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543017 Supplier Invitrogen™ Supplier No. MA543017
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human SETD7/SET7/9

Histone methyltransferase that specifically monomethylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates 'Lys-189' of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates 'Lys-372' of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD7
Applications ELISA
Classification Monoclonal
Clone 7C1B7E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Setd7
Gene Accession No. Q8WTS6
Gene Alias 1600028F23Rik; FLJ21193; H3-K4 methyltransferase; H3-K4-HMTase; H3-K4-HMTase SETD7; H3K4MT; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; histone-lysine N-methyltransferase SETD7; Kiaa1717; KMT7; lysine N-methyltransferase 7; mKIAA1717; OTTHUMP00000220049; SET domain containing (lysine methyltransferase) 7; SET domain containing 7, histone lysine methyltransferase; SET domain containing 7, lysine methyltransferase; SET domain containing lysine methyltransferase 7; SET domain-containing protein 7; Set7; Set7/9; SET9; Setd7
Gene Symbols Setd7
Host Species Mouse
Immunogen Recombinant Human SETD7 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80854
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.