Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7 Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503354S


Description
SETD7 Monoclonal antibody specifically detects SETD7 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
SETD7 | |
Monoclonal | |
0.25 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9ERR1, Q9GZM8 | |
Mouse | |
Affinity chromatography | |
RUO | |
81565, 83431 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI5B11 | |
Unconjugated | |
Ndel1 | |
1600028F23Rik; FLJ21193; H3-K4 methyltransferase; H3-K4-HMTase; H3-K4-HMTase SETD7; H3K4MT; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; histone-lysine N-methyltransferase SETD7; Kiaa1717; KMT7; lysine N-methyltransferase 7; mKIAA1717; OTTHUMP00000220049; SET domain containing (lysine methyltransferase) 7; SET domain containing 7, histone lysine methyltransferase; SET domain containing 7, lysine methyltransferase; SET domain containing lysine methyltransferase 7; SET domain-containing protein 7; Set7; Set7/9; SET9; Setd7 | |
Full length human recombinant protein of human NDEL1 produced in HEk293 cell. | |
30 μL | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction