Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD7 Mouse anti-Human, Mouse, Clone: OTI2E3, lyophilized, TrueMAB™
SDP

Catalog No. 501662089
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-2089 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-2089 Supplier OriGene Supplier No. CF503377
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Histone methyltransferase that specifically monomethylates ′Lys-4′ of histone H3. H3 ′Lys-4′ methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates ′Lys-189′ of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates ′Lys-372′ of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. Also able to demethylated ′Lys-372′ of p53/TP53 in vitro.

Specifications

Antigen SETD7
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2E3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SETD7
Gene Accession No. Q8VHL1, Q8WTS6
Gene Alias KMT7, SET7, SET7/9, SET9
Gene Symbols SETD7
Host Species Mouse
Immunogen Full length human recombit protein of human SETD7 produced in HEK293 cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73251, 80854
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.