Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530920 Supplier Invitrogen™ Supplier No. PA530920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HCT116. Predicted reactivity: Mouse (96%), Rat (97%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Histone methyltransferase that specifically monomethylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates 'Lys-189' of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates 'Lys-372' of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Setd7
Gene Accession No. Q8WTS6
Gene Alias 1600028F23Rik; FLJ21193; H3-K4 methyltransferase; H3-K4-HMTase; H3-K4-HMTase SETD7; H3K4MT; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; histone-lysine N-methyltransferase SETD7; Kiaa1717; KMT7; lysine N-methyltransferase 7; mKIAA1717; OTTHUMP00000220049; SET domain containing (lysine methyltransferase) 7; SET domain containing 7, histone lysine methyltransferase; SET domain containing 7, lysine methyltransferase; SET domain containing lysine methyltransferase 7; SET domain-containing protein 7; Set7; Set7/9; SET9; Setd7
Gene Symbols Setd7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SETD7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.