Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD8 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299864V
Description
SETD8 Polyclonal specifically detects SETD8 in Human samples. It is validated for Western Blot, ELISA.Specifications
SETD8 | |
Polyclonal | |
Western Blot, ELISA | |
H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8 | |
Produced in rabbits immunized with purified, recombinant SETD8 (NP_065115.3; Lys195-His352). | |
0.1 mL | |
Cell Cycle and Replication, Epigenetics | |
387893 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction