Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD8 Monoclonal Antibody (OTI2H5), TrueMAB™, OriGene
SDP

Catalog No. 501681977
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1977 Supplier OriGene Supplier No. TA808218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone methyltransferase that specifically monomethylates ′Lys-20′ of histone H4. H4 ′Lys-20′ monomethylation is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. Mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. Required for cell proliferation, probably by contributing to the maintece of proper higher order structure of DNA during mitosis. Involved in chromosome condensation and proper cytokinesis. Nucleosomes are preferred as substrate compared to free histones.

Specifications

Antigen SETD8
Applications Western Blot
Classification Monoclonal
Clone OTI2H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KMT5A
Gene Accession No. Q9NQR1
Gene Alias PR-Set7, SET07, SET8, SETD8
Gene Symbols KMT5A
Host Species Mouse
Immunogen Full length human recombit protein of human SETD8 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387893
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.