Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETDB1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079776AF532
Description
SETDB1 Polyclonal antibody specifically detects SETDB1 in Human samples. It is validated for Western BlotSpecifications
SETDB1 | |
Polyclonal | |
Western Blot | |
ERG-associated protein with SET domain, ESETEC 2.1.1.43, H3-K9-HMTase 4, Histone H3-K9 methyltransferase 4, histone-lysine N-methyltransferase SETDB1, histone-lysine N-methyltransferase, H3lysine-9 specific 4, KG1T, KIAA0067H3-K9-HMTase4, KMT1EERG-associated protein with a SET domain, ESET, Lysine N-methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1 | |
The immunogen recognized by this antibody maps to a region between residues 1125 and 1175 of human SET Domain, Bifurcated 1 using the numbering given in entry NP_036564.2 (GeneID 9869). | |
0.1 mL | |
Epigenetics, Transcription Factors and Regulators | |
9869 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction