Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETDB1 Mouse anti-Human, Clone: 1H6E5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SETDB1 |
---|---|
Clone | 1H6E5 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6725
![]() ![]() |
Proteintech Group Inc
662931IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N (6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.Specifications
SETDB1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ESET, H3 K9 HMTase 4, H3 K9 HMTase4, KG1T, KIAA0067, KMT1E, Lysine N methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1, SETDB1 | |
SETDB1 | |
IgG1 | |
Protein G | |
Antibody |
1H6E5 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q15047 | |
9869 | |
SETDB1 Fusion Protein Ag21644 | |
Primary | |
-20°C | |
SETDB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title