Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETDB1, Mouse anti-Mouse, Clone: F0838, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $498.79
Specifications
Antigen | SETDB1 |
---|---|
Clone | F0838 |
Dilution | Western Blotting Analysis: A representative lot detected SETDB1 in Western Blotting applications (Matsumura, Y, et. al. (2015). Mol Cell. 60(4):584-96). |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE106625
|
MilliporeSigma
MABE106625UG |
25 μg |
Each for $192.34
|
|
|||||
MABE1066100
|
MilliporeSigma
MABE1066100UG |
100 μg |
Each for $498.79
|
|
|||||
Description
Anti-SETDB1, clone F0838, Cat. No. MABE1066, is a highly specific mouse monoclonal antibody that detects SETDB1 and has been tested for use in Western Blotting.
Histone-lysine N-methyltransferase SETDB1 (UniProt: O88974; also known as EC: 2.1.1.43, ERG-associated protein with SET domain, ESET, SET domain bifurcated 1) is encoded by the Setdb1 (also known as Eset, Kiaa0067) gene in human. SETDB1 is ubiquitously expressed, but a strong expression is observed in liver and testis. It is a histone methyltransferase that specifically trimethylates Histone 3 Lysine 9 (H3K9). H3K9 trimethylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 (CBX1, CBX3 and/or CBX5) proteins to methylated histones. It mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. SETDB1 is reported to inhibit adipogenesis. It has also been shown that H3K4/H3K9me3 bivalent chromatin signature keeps expression of Cebpa and Pparg genes very low. Depletion of SETDB1 can induce adipogenesis in mesenchymal stem cells and in preadipocytes. SETDB1 contains two Tudor domains (aa 257-320 and 347-403) and one MBD domain (aa 611-682). It also has a SET domain (aa 820-1282), a pre-SET (aa 744-817), and a post-SET domain (aa 1291-1307). The pre-SET, SET and post-SET domains are required for its methyltransferase activity. In the pre-SET domain, Cys residues bind 3 zinc ions that are arranged in a triangular cluster; some of these Cys residues contribute to the binding of two zinc ions within the cluster. (Ref.: Matsumura, Y., et al., (2015), Mol. Cell 60(4); 584- 596).Specifications
SETDB1 | |
Western Blotting Analysis: A representative lot detected SETDB1 in Western Blotting applications (Matsumura, Y, et. al. (2015). Mol Cell. 60(4):584-96). | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Histone-lysine N-methyltransferase SETDB1;EC: 2.1.1.43;ERG-associated protein with SET domain;ESET;SET domain bifurcated 1 | |
His-tagged recombinant fragment corresponding to 213 amino acids from the N-terminal half of murine Histone-lysine N-methyltransferase SETDB1. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
F0838 | |
Western Blot | |
Unconjugated | |
Mouse | |
Epigenetics & Nuclear Function | |
Purified mouse monoclonal antibody IgG1 in PBS with 0.01% sodium azide. | |
Setdb1;Eset;Kiaa0067 | |
IgG1 κ | |
Protein G purified | |
Clone F0838 recognizes SETDB1 in murine embryonic stem cells. It targets an epitope with in 213 amino acids from the N-terminal half of isoform 1 and 7. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title