Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETDB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142718
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142718 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142718 Supplier Invitrogen™ Supplier No. PA5142718
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_114121.2; NP_001153780.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETDB2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SETDB2
Gene Accession No. Q96T68
Gene Alias bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; Clld8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; Setdb2; Setdb2-Phf11-1-Phf11-2 fusion
Gene Symbols SETDB2
Host Species Goat
Immunogen Peptide with sequence C-QRRHAKTNSHSSALH (aa 174-188).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83852
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.