Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETDB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100857 Supplier Invitrogen™ Supplier No. PA5100857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SETDB2.

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETDB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SETDB2
Gene Accession No. Q8C267, Q96T68
Gene Alias bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; Clld8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; Setdb2; Setdb2-Phf11-1-Phf11-2 fusion
Gene Symbols SETDB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SETDB2(Accession Q96T68), corresponding to amino acid residues M1-Q33.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239122, 83852
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.