Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETDB2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518112
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).
Specifications
SETDB2 | |
Polyclonal | |
Unconjugated | |
SETDB2 | |
bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; Clld8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; Setdb2; Setdb2-Phf11-1-Phf11-2 fusion | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
83852 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96T68 | |
SETDB2 | |
Synthetic peptide sequence (GEKNGDAKTFWME) corresponding to the N-terminus amino acids of SETDB2 (aa 2-15). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction