Learn More
Invitrogen™ SETDB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110357
Description
Immunogen sequence: SGACLMKMPL NLKGENPLQL PIKCHFQRRH AKTNSHSSAL HVSYKTPCGR SLRNVEEVFR YLLETECNFL FTDNFSFNTY VQLARNYPKQ KEVVSDVDIS NGVESVPISF CNEIDSRKLP QFKYRKTVWP RAYNLTNFSS MFTDSCDCSE GCIDITKCAC LQLTARNAKT SPLSSDKITT GYKYKRLQRQ IPTGIYECSL LCKCNRQLCQ NRVVQHGPQV RLQVFKTEQK G.
Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).
Specifications
SETDB2 | |
Polyclonal | |
Unconjugated | |
SETDB2 | |
bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; Clld8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; Setdb2; Setdb2-Phf11-1-Phf11-2 fusion | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100361710, 239122 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.04 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q8C267 | |
SETDB2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 148-378 of human SETDB2 (NP_1141212). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.