Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Setdb2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990930
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0930 25 μL
50-199-0929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0930 Supplier Rockland Immunochemicals Supplier No. 200401N99S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This Protein A purified antibody is directed against mouse Setdb2. Expect reactivity with Setdb2 from mouse and human sources. Reactivity against homologues from other sources is not known.

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).

Specifications

Antigen Setdb2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene SETDB2
Gene Accession No. Q8C267, Q96T68
Gene Alias bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; CLLD8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; SETDB2; Setdb2-Phf11-1-Phf11-2 fusion
Gene Symbols SETDB2
Host Species Rabbit
Immunogen Setdb2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a full length protein corresponding to mouse Setdb2.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239122, 83852
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.