Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETMAR Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197739AF750
Description
SETMAR Polyclonal antibody specifically detects SETMAR in Human samples. It is validated for Western BlotSpecifications
SETMAR | |
Polyclonal | |
Western Blot | |
EC 2.1.1.43, histone-lysine N-methyltransferase SETMAR, hsMar1, metnase, SET domain and mariner transposase fusion gene, SET domain and mariner transposase fusion gene-containing protein | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human SET Domain and Mariner Transposase Fusion Gene-Containing Protein using the numbering given in entry BAD96454.1 (GeneID 6419). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6419 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction