Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SETMAR Polyclonal Antibody

Catalog No. PI720343
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720343 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720343 Supplier Invitrogen™ Supplier No. 720343

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Bovine, Sheep.

SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETMAR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene SETMAR
Gene Accession No. Q53H47, Q80UJ9
Gene Alias 5830404F24Rik; epidermal tumor expressed transcript 2; Etet2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Mar1; METNASE; SET domain and mariner transposase fusion gene; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain and mariner transposase fusion protein homolog; SET domain without mariner transposase fusion; SET domain without mariner transposase fusion protein; Setmar; Transposon Hsmar1 transposase
Gene Symbols SETMAR
Host Species Rabbit
Immunogen Protein corresponding to human SETMAR [aa15-aa303].
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6419, 74729
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.