Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETMAR Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518116
Description
This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.
Specifications
SETMAR | |
Polyclonal | |
Unconjugated | |
SETMAR | |
5830404F24Rik; epidermal tumor expressed transcript 2; Etet2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Mar1; METNASE; SET domain and mariner transposase fusion gene; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain and mariner transposase fusion protein homolog; SET domain without mariner transposase fusion; SET domain without mariner transposase fusion protein; Setmar; Transposon Hsmar1 transposase | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
6419 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q53H47 | |
SETMAR | |
Synthetic peptide sequence (RWQKCVDCNGSYFD) corresponding to the C-terminus amino acids of SETMAR (aa 671-684). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction