Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETX Recombinant Rabbit Monoclonal Antibody (BLR050F)

Rabbit Recombinant Monoclonal Antibody
$173.10 - $452.00
Specifications
| Antigen | SETX |
|---|---|
| Clone | BLR050F |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544293
![]() |
Invitrogen™
MA544293 |
100 μL |
Each for $452.00
|
|
|||||
|
PIMA544294
|
Invitrogen™
MA544294 |
20 μL |
Each for $173.10
|
|
|||||
Description
SETX Recombinant Monoclonal Antibody for Western Blot, IP
Probable RNA/DNA helicase involved in diverse aspects of RNA metabolism and genomic integrity. Plays a role in transcription regulation by its ability to modulate RNA Polymerase II (Pol II) binding to chromatin and through its interaction with proteins involved in transcription (PubMed:19515850, PubMed:21700224). Contributes to the mRNA splicing efficiency and splice site selection (PubMed:19515850). Required for the resolution of R-loop RNA-DNA hybrid formation at G-rich pause sites located downstream of the poly(A) site, allowing XRN2 recruitment and XRN2-mediated degradation of the downstream cleaved RNA and hence efficient RNA polymerase II (RNAp II) transcription termination (PubMed:19515850, PubMed:21700224, PubMed:26700805). Required for the 3' transcriptional termination of PER1 and CRY2, thus playing an important role in the circadian rhythm regulation. Involved in DNA double-strand breaks damage response generated by oxidative stress (PubMed:17562789). In association with RRP45, targets the RNA exosome complex to sites of transcription-induced DNA damage (PubMed:24105744). Plays a role in the development and maturation of germ cells: essential for male meiosis, acting at the interface of transcription and meiotic recombination, and in the process of gene silencing during meiotic sex chromosome inactivation (MSCI). May be involved in telomeric stability through the regulation of telomere repeat-containing RNA (TERRA) transcription (PubMed:21112256). Plays a role in neurite outgrowth in hippocampal cells through FGF8-activated signaling pathways. Inhibits retinoic acid-induced apoptosis (PubMed:21576111).Specifications
| SETX | |
| 1 mg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| Q7Z333 | |
| 23064 | |
| Peptide representing a region between residues 900 and 950 of human Senataxin. | |
| Antibody |
| BLR050F | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| SETX | |
| A130090N03; A930037J23Rik; Als4; amyotrophic lateral sclerosis 4 homolog; amyotrophic lateral sclerosis 4 protein; Amyotrophic lateral sclerosis 4 protein homolog; AOA2; AW060766; bA479K20.2; DEAxQ-box helicase; KIAA0625; mKIAA0625; Probable helicase senataxin; RGD1565575; SCAR1; Sen1; SEN1 homolog; senataxin; Setx; tRNA splicing endonuclease regulator 1 | |
| SETX | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title