Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEZ6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552854
Description
Immunogen sequence: PGDVEHSRRL ISSPKFPVGA TVQYICDQGF VLMGSSILTC HDRQAGSPKW SDRAPKCLLE QLKPCHGLSA PENGARSPEK QLHPAGATIH FSCAPGYVLK GQASIKCVPG HPSHWSDPPP ICRA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Sez6 may play a role in cell-cell recognition and in neuronal membrane signaling. Seems to be important for the achievement of the necessary balance between dendrite elongation and branching during the elaboration of a complex dendritic arbor. Involved in the development of appropriate excitatory synaptic connectivity.
Specifications
SEZ6 | |
Polyclonal | |
Unconjugated | |
Sez6 | |
brain-specific receptor-like protein C; BSRPC; BSRP-C; D11Bhm177e; hSEZ-6; Kiaa4158; Seizure protein 6; seizure protein 6 homolog; seizure related 6 homolog; seizure related 6 homolog (mouse); seizure related gene 6; SEZ6; SEZ-6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53EL9 | |
Sez6 | |
Recombinant protein corresponding to Human SEZ6. Recombinant protein control fragment (Product #RP-89899). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction