Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEZ6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547683
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547683 Supplier Invitrogen™ Supplier No. PA547683
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse (rm) BSRP-A and rmBSRP-B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Sez6 may play a role in cell-cell recognition and in neuronal membrane signaling. Seems to be important for the achievement of the necessary balance between dendrite elongation and branching during the elaboration of a complex dendritic arbor. Involved in the development of appropriate excitatory synaptic connectivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEZ6
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Sez6
Gene Accession No. Q7TSK2
Gene Alias brain-specific receptor-like protein C; BSRPC; BSRP-C; D11Bhm177e; hSEZ-6; Kiaa4158; Seizure protein 6; seizure protein 6 homolog; seizure related 6 homolog; seizure related 6 homolog (mouse); seizure related gene 6; SEZ6; SEZ-6
Gene Symbols Sez6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SEZ6/BSRP-C Gly19-Asn902.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20370
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.