Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEZ6L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550757 Supplier Thermo Scientific Supplier No. PA550757
Add to Cart
Add to Cart

SEZ6L Polyclonal Antibody

SEZ6L Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SEZ6L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SEZ6L
Gene Accession No. Q6P1D5, Q9BYH1
Gene Alias Acig1; acupuncture induced gene 1; acupuncture-induced protein 1-L; AI843918; AIG1; Aig1l; AW539724; Brain-specific receptor-like protein B; BSRP-B; KIAA0927; mKIAA0927; RGD1563628; seizure 6-like protein; seizure related 6 homolog (mouse)-like; seizure related 6 homolog like; seizure related gene 6-like; Sez6l; UNQ2542/PRO6094
Gene Symbols SEZ6L
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human seizure related 6 homolog (mouse)-like.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23544, 56747
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.