Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEZ6L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987047
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7047 Supplier Bioss Supplier No. BS6103R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEZ6L (seizure related 6 homolog-like) is a 1,024 amino acid single-pass type I membrane protein that localizes to the endoplasmic reticulum. Widely expressed, SEZ6L is a candidate tumor suppressor gene and may contribute to specialized endoplasmic reticulum functions in neurons. SEZ6L contains three CUB domains and five sushi (CCP/SCR) domains, which may be involved in protein-protein interactions and signal transduction. The gene encoding SEZ6L is located on human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.

Specifications

Antigen SEZ6L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEZ6L
Gene Accession No. Q9BYH1
Gene Alias Acig1; acupuncture induced gene 1; Acupuncture-induced protein 1-L; AI843918; AIG1; Aig1l; AW539724; Brain-specific receptor-like protein B; BSRP-B; Kiaa0927; mKIAA0927; Seizure 6-like protein; seizure related 6 homolog (mouse)-like; seizure related 6 homolog like; seizure related gene 6-like; SEZ6L; UNQ2542/PRO6094
Gene Symbols SEZ6L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SEZ6L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23544
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.