Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEZ6L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564172 Supplier Invitrogen™ Supplier No. PA564172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGLISDAQSL YVELLSETPA NPLLLSLRFE AFEEDRCFAP FLAHGNVTTT DPEYRPGALA TFSCLPGYAL EPPGPPNAIE CV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 99%.

The SEZ6L2 gene encodes the protein seizure-related 6 homolog-like 2, which plays roles in neural development and synaptic function. SEZ6L2 is part of the SEZ6 protein family recognized for their participation in the formation and maintenance of neural connections. This protein is prominently expressed in the brain and is involved in the molecular processes underlying synaptic differentiation and dendritic growth, crucial for cognitive functions and neurodevelopment. SEZ6L2 interacts with other neural proteins to mediate signaling pathways that regulate the balance between excitation and inhibition in neural circuits, which is vital for proper neurological function and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEZ6L2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEZ6L2
Gene Accession No. Q6UXD5
Gene Alias AI835913; AW121566; Brain-specific receptor-like protein A; BSRPA; BSRP-A; PSK; Psk1; PSK-1; seizure 6-like protein 2; seizure related 6 homolog (mouse)-like 2; seizure related 6 homolog like 2; seizure related 6 homolog-like 2; seizure related 6-like protein 2; SEZ6L2; type I transmembrane receptor (seizure-related protein); UNQ1903/PRO4349
Gene Symbols SEZ6L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEZ6L2. Recombinant protein control fragment (Product #RP-103562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26470
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.