Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299041AF488
Description
SF-1/NR5A1/Steroidogenic Factor 1 Polyclonal specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human samples. It is validated for Western Blot.Specifications
SF-1/NR5A1/Steroidogenic Factor 1 | |
Polyclonal | |
Western Blot | |
AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human SF-1/NR5A1/Steroidogenic Factor 1. | |
0.1 mL | |
GPCR | |
2516 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction