Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SF-1/NR5A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | SF-1/NR5A1/Steroidogenic Factor 1 |
---|---|
Clone | 1036724 |
Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB107241SP
![]() |
R&D Systems
MAB107241SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10724110
![]() |
R&D Systems
MAB107241100 |
100 μg |
Each for $432.50
|
|
|||||
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
SF-1/NR5A1/Steroidogenic Factor 1 | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AD4BP, AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, NR5A1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
Human SF-1/NR5A1 synthetic peptide, Accession # Q13286 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1036724 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q13286 | |
2516 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title